Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173024.3 | NP_766612.1 | 2 | 202 | ||
GenBank mRNA | AK037160.1 | - | 2 | 204 | ||
BC125418.1 | - | 2 | 185 | 40129933 | ||
BC145808.1 | - | 2 | 185 | 40131111 |