Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC024139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142968.1 | NP_001136440.1 | 3 | 433 | ||
GenBank mRNA | AK078032.1 | - | 3 | 421 | ||
AK081549.1 | - | 3 | 435 | |||
BC060242.1 | - | 3 | 473 |