Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252067.1 | NP_001238996.1 | 2 | 126 | ||
NM_173048.3 | NP_766636.2 | 2 | 126 | |||
GenBank mRNA | AK031086.1 | - | 2 | 98 | ||
AK082327.1 | - | 2 | 90 | |||
AK122212.2 | - | 2 | 95 | |||
AK132183.1 | - | 2 | 94 | |||
AK147882.1 | - | 2 | 95 | |||
BC024266.1 | - | 733 | ||||
BC056424.1 | - | 2 | 80 | |||
BC085617.1 | - | 2 | 105 | |||
BC110676.1 | - | 2 | 82 | |||
BY268727.1 | - | 2 | 126 | |||
BY296723.1 | - | 2 | 117 |