Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167923.1 | NP_001161395.1 | 3 | 490 | ||
GenBank mRNA | AM748256.1 | - | 3 | 490 |
Gene Symbol: Col6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102607.1 | NP_001096077.1 | 3 | 572 | ||
NM_172927.3 | NP_766515.2 | 3 | 607 | |||
GenBank mRNA | AK054356.1 | - | 3 | 608 | ||
AK087828.1 | - | 3 | 648 | |||
AM748259.1 | - | 3 | 572 | |||
AM748260.1 | - | 1 | 292 | |||
BC150695.1 | - | 2 | 391 | 9087606 |