Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114098.1 | NP_001107570.1 | 7 | 3480 | ||
NM_172963.4 | NP_766551.3 | 7 | 3478 | |||
GenBank mRNA | AK051750.1 | - | 2 | 1212 | ||
AK147205.1 | - | 7 | 3480 | |||
AK147631.1 | - | 7 | 3472 | |||
AK147691.1 | - | 7 | 3472 | |||
BC032156.2 | - | 3 | 600 | |||
BC060225.1 | - | 5 | 1726 | 30356997 |