Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172964.4 | NP_766552.3 | 2, 3 | 449 | ||
GenBank mRNA | AK087523.1 | - | 2, 3 | 462 | ||
AK133720.1 | - | 2, 3 | 291 | |||
AK141901.1 | - | 2, 3 | 399 | |||
AK173147.1 | - | 1, 2 | 76 | |||
BC066788.1 | - | 2, 3 | 439 | 30049271 |