Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172974.2 | NP_766562.1 | 3 | 332 | ||
GenBank mRNA | AF071317.1 | - | 3 | 310 | ||
AK053615.1 | - | 3 | 291 | |||
AK081309.1 | - | 3 | 332 | |||
AK168740.1 | - | 3 | 291 | |||
AK170140.1 | - | 3 | 291 |