Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290786.1 | NP_001277715.1 | 2 | 649 | ||
NM_172979.3 | NP_766567.1 | 2 | 649 | |||
GenBank mRNA | AK052840.1 | - | 2 | 536 | ||
AK142590.1 | - | 2 | 536 | |||
AY198336.1 | - | 2 | 565 | |||
AY198337.1 | - | 2 | 532 | |||
BC116796.1 | - | 2 | 528 | 40126115 | ||
BC119176.1 | - | 2 | 528 | 8733925 |