Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172988.4 | NP_766576.1 | 3 | 412 | ||
GenBank mRNA | AK028786.1 | - | 3 | 396 | ||
AK032391.1 | - | 3 | 391 | |||
AK051429.1 | - | 3 | 363 | |||
AK083661.1 | - | 3 | 386 | |||
BC033265.1 | - | 3 | 339 | 3983942 | ||
BY128470.1 | - | 3 | 276 |