Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172992.3 | NP_766580.3 | 4, 5 | 384 | ||
GenBank mRNA | AK035536.1 | - | 4, 5 | 338 | ||
AK037021.1 | - | 4, 5 | 271 | |||
AK042382.1 | - | 4, 5 | 363 | |||
AK042790.1 | - | 4, 5 | 306 | |||
AK152951.1 | - | 4, 5 | 337 | |||
BC052841.1 | - | 4, 5 | 297 | 30050876 | ||
CB245749.1 | - | 4, 5 | 384 |