Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109749.1 | NP_001103219.1 | 6 | 665 | ||
NM_001109751.1 | NP_001103221.1 | 5 | 753 | |||
NM_173004.3 | NP_766592.2 | 6 | 665 | |||
GenBank mRNA | AK036262.1 | - | 6 | 635 | ||
AK043507.1 | - | 3 | 610 | |||
AK087738.1 | - | 4 | 379 | |||
AK163520.1 | - | 3 | 458 | |||
BC046975.1 | - | 3 | 287 | |||
BC115766.1 | - | 6 | 555 | 40060288 |