Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173008.2 | NP_766596.1 | 7 | 1245 | ||
GenBank mRNA | AK079906.1 | - | 7 | 1246 | ||
AK165666.1 | - | 7 | 1307 | |||
BC141044.1 | - | 7 | 1243 | 9055656 | ||
EU850434.1 | - | 7 | 1246 |