Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173016.3 | NP_766604.2 | 4, 5 | 789 | ||
GenBank mRNA | AK122527.1 | - | 4, 5 | 789 | ||
AK189227.1 | - | 1, 2 | 15 | |||
AK199051.1 | - | 1, 2 | 15 | |||
BC056927.1 | - | 4, 5 | 799 | 5719211 |