Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173052.2 | NP_766640.1 | 4, 5 | 555 | ||
GenBank mRNA | AF426025.1 | - | 3, 4 | 438 | ||
AK018226.1 | - | 4, 5 | 534 | |||
BC030354.1 | - | 4, 5 | 500 | 5367365 |