Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc2hc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173181.3 | NP_775273.1 | 3, 4 | 303 | ||
GenBank mRNA | AK014202.1 | - | 3, 4 | 239 | ||
AK083941.1 | - | 3, 4 | 247 | |||
AK085187.1 | - | 3, 4 | 240 | |||
AK170609.1 | - | 3, 4 | 244 | |||
BC044809.1 | - | 3, 4 | 225 | |||
BC087731.1 | - | 267 | 6850296 | |||
BY131077.1 | - | 3, 4 | 306 |