Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172914.2 | NP_766502.1 | 5 | 679 | ||
GenBank mRNA | AK077130.1 | - | 5 | 680 | ||
BC060957.1 | - | 5 | 677 | 6705542 |