Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172965.2 | NP_766553.1 | 19 | 2938 | ||
GenBank mRNA | AK011978.1 | - | 3 | 236 | ||
AK053675.1 | - | 19 | 2939 | |||
BC063075.1 | - | 16 | 2465 | 5721433 | ||
BC066030.1 | - | 19 | 2953 |