Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172980.2 | NP_766568.1 | 9 | 2069 | ||
GenBank mRNA | AF079853.2 | - | 8 | 944 | ||
AK080957.1 | - | 9 | 2070 | |||
AK152776.1 | - | 8 | 1004 |
Gene Symbol: Gm14085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085518.1 | NP_001078987.1 | 8 | 1907 |