Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039866.1 | - | 9, 10 | 1156 | ||
AK050894.1 | - | 12, 13 | 1598 | |||
BC012520.1 | - | 1, 2 | 101 | |||
BC019659.1 | - | 3, 4 | 462 | |||
BC050089.1 | - | 12, 13 | 1596 | 30018732 |