Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172991.4 | NP_766579.3 | 4 | 969 | ||
GenBank mRNA | AK047811.1 | - | 4 | 501 | ||
BC029549.1 | - | 4 | 833 | 5012510 |