Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163.3 | NP_001154.2 | 2 | 1088 | ||
XM_005251968.3 | XP_005252025.1 | 2 | 1119 | |||
XM_011518617.2 | XP_011516919.1 | 2 | 1043 | |||
XM_017014670.1 | XP_016870159.1 | 2 | 1332 | |||
GenBank mRNA | AF029106.1 | - | 2 | 1010 | ||
AF047347.1 | - | 2 | 1051 | |||
L04953.1 | - | 1 | 425 |