Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC048403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173022.2 | NP_766610.2 | 1 | 149 | ||
GenBank mRNA | AK087110.1 | - | 1 | 137 | ||
AK138662.1 | - | 1 | 139 | |||
AK162911.1 | - | 1 | 139 | |||
AK166238.1 | - | 1 | 148 | |||
AK169891.1 | - | 1 | 139 | |||
BC065058.1 | - | 1 | 149 | 5720152 |