Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp8nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173031.3 | NP_766619.2 | 10 | 1646 | ||
GenBank mRNA | AK028610.1 | - | 10 | 1648 | ||
BC066135.1 | - | 8 | 781 | |||
BC150940.1 | - | 10 | 1601 | |||
BC172093.1 | - | 10 | 1601 |