Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030055.1 | NP_001025226.1 | 2 | 513 | ||
NM_001173.2 | NP_001164.2 | 2 | 513 | |||
XM_005267635.3 | XP_005267692.1 | 2 | 482 | |||
XM_005267636.3 | XP_005267693.1 | 2 | 728 | |||
XM_017021287.1 | XP_016876776.1 | 2 | 432 | |||
XM_017021288.1 | XP_016876777.1 | 2 | 482 | |||
GenBank mRNA | AK292966.1 | - | 1 | 378 | ||
BC032723.2 | - | 1 | 367 | |||
BC050059.1 | - | 2 | 439 | 5296634 | ||
U17032.1 | - | 2 | 500 |