Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173180.3 | NP_775272.1 | 13 | 2140 | ||
GenBank mRNA | AK004549.1 | - | 13 | 2140 | ||
AK075965.1 | - | 13 | 2140 | |||
AK147110.1 | - | 13 | 2143 | |||
AK158990.1 | - | 13 | 2108 | |||
AK167975.1 | - | 13 | 2139 | |||
AK169325.1 | - | 13 | 2135 | |||
AK169342.1 | - | 13 | 2135 |