Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173185.2 | NP_775277.2 | 14 | 2094 | ||
GenBank mRNA | AK090262.1 | - | 14 | 2095 | ||
BC064645.1 | - | 13 | 2023 | 5363779 |