Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001175.6 | NP_001166.3 | 4 | 542 | ||
NM_001321420.1 | NP_001308349.1 | 5 | 700 | |||
NM_001321421.1 | NP_001308350.1 | 4 | 421 | |||
NM_001321422.1 | NP_001308351.1 | 4 | 543 | |||
NM_001321423.1 | NP_001308352.1 | 4 | 409 | |||
NR_135637.1 | 2 | 128 | ||||
GenBank mRNA | AA143425.1 | - | 4 | 353 | ||
AB451315.1 | - | 3 | 296 | |||
AB451445.1 | - | 3 | 296 | |||
AF498927.1 | - | 3 | 296 | |||
AK311807.1 | - | 4 | 372 | |||
AV717900.1 | - | 4 | 420 | |||
BC009200.2 | - | 4 | 346 | 3621138 | ||
BE744996.1 | - | 4 | 407 | |||
BG390701.1 | - | 5 | 517 | |||
BI762648.1 | - | 4 | 541 | |||
BP365648.1 | - | 2 | 128 | |||
BY996333.1 | - | 4 | 406 | |||
L07916.1 | - | 4 | 370 | |||
L20688.1 | - | 4 | 448 | |||
X69549.1 | - | 4 | 350 |