Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173364.5 | NP_775540.4 | 7 | 2097 | ||
GenBank mRNA | AK036132.1 | - | 1 | 260 | ||
AY341877.1 | - | 6 | 1938 | |||
BC027167.1 | - | 1 | 843 | |||
BC034572.1 | - | 1 | 707 | |||
BC055684.1 | - | 1 | 394 | |||
BC099896.1 | - | 6 | 1909 | 30545532 | ||
BC099930.1 | - | 7 | 2042 | 30058096 |