Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128170.2 | NP_001121642.1 | 2 | 360 | ||
NM_001128171.2 | NP_001121643.1 | 3 | 494 | |||
NM_001276279.1 | NP_001263208.1 | 3 | 491 | |||
NM_173369.3 | NP_775545.1 | 2 | 360 | |||
GenBank mRNA | AK039054.1 | - | 3 | 489 | ||
AK042764.1 | - | 2 | 327 | |||
AK122389.1 | - | 3 | 475 | |||
AK133479.1 | - | 2 | 324 | |||
AK147204.1 | - | 2 | 329 | |||
AK147426.1 | - | 2 | 324 | |||
AK167424.1 | - | 3 | 455 | |||
BC042438.1 | - | 2 | 313 | 3983771 |