Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC068281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170858.1 | NP_001164329.1 | 2 | 343 | ||
NM_173416.3 | NP_775592.2 | 2 | 343 | |||
GenBank mRNA | AK040407.1 | - | 2 | 300 | ||
AK155130.1 | - | 2 | 263 | |||
AK164024.1 | - | 3 | 504 | |||
AK170427.1 | - | 2 | 242 | |||
BC068281.1 | - | 2 | 280 | 30008957 |