Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pskh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173432.2 | NP_775608.1 | 2 | 640 | ||
GenBank mRNA | AK053397.1 | - | 2 | 674 | ||
AK153861.1 | - | 2 | 687 | |||
AK154570.1 | - | 2 | 670 | |||
AK218640.1 | - | 1 | 164 | |||
BC050128.1 | - | 2 | 640 | 30024345 |