Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142610.1 | NP_001136082.1 | 3, 4 | 742 | ||
NM_014683.3 | NP_055498.3 | 3, 4 | 742 | |||
XM_017025424.1 | XP_016880913.1 | 3, 4 | 567 | |||
XM_017025425.1 | XP_016880914.1 | 3, 4 | 567 | |||
XM_017025426.1 | XP_016880915.1 | 3, 4 | 567 | |||
XM_017025427.1 | XP_016880916.1 | 2, 3 | 143 | |||
XM_017025428.1 | XP_016880917.1 | 2, 3 | 143 | |||
GenBank mRNA | AB014523.1 | - | 3, 4 | 733 | ||
AK098316.1 | - | 2, 3 | 367 | |||
AK314345.1 | - | 3, 4 | 267 | |||
BC034988.1 | - | 4, 5 | 307 | 5268025 |