Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl14ep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025102.1 | NP_001020273.1 | 2 | 275 | ||
NM_173750.2 | NP_776111.1 | 2 | 229 | |||
GenBank mRNA | AK012215.1 | - | 2 | 273 | ||
AK076781.1 | - | 2 | 230 | |||
AK134965.1 | - | 2 | 266 | |||
AK165082.1 | - | 2 | 274 | |||
BB871621.1 | - | 2 | 275 | |||
BC054106.1 | - | 2 | 261 | 30021622 |