Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilvbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173751.4 | NP_776112.1 | 2, 3 | 319 | ||
GenBank mRNA | AK087956.1 | - | 1, 2 | 193 | ||
AK133553.1 | - | 1, 2 | 249 | |||
AK138031.1 | - | 1, 2 | 168 | |||
AK143918.1 | - | 1, 2 | 218 | |||
AK146075.1 | - | 1, 2 | 247 | |||
AK169899.1 | - | 1, 2 | 245 | |||
AK196554.1 | - | 1, 2 | 115 | |||
BC052054.1 | - | 1, 2 | 653 | |||
BC056459.1 | - | 2, 3 | 277 | 5718157 | ||
CD768274.1 | - | 2, 3 | 323 |