Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173365.2 | NP_775541.1 | 2 | 1313 | ||
GenBank mRNA | AK040547.1 | - | 2 | 1314 | ||
BC117783.2 | - | 2 | 1175 | 40092101 |