Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173372.2 | NP_775548.2 | 7 | 1329 | ||
GenBank mRNA | BC034118.1 | - | 3 | 330 | ||
BC041684.1 | - | 2 | 338 | 4502870 | ||
BC152480.1 | - | 6 | 1109 |
Set Membership: |
![]() |