Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291124.1 | NP_001278053.1 | 2 | 421 | ||
NM_173396.3 | NP_775572.1 | 2 | 421 | |||
GenBank mRNA | AK017884.1 | - | 2 | 289 | ||
AK029490.1 | - | 2 | 351 | |||
AK157623.1 | - | 2 | 329 | |||
AK178732.1 | - | 1 | 35 | |||
AK185386.1 | - | 1 | 56 | |||
AK186516.1 | - | 1 | 35 | |||
AK207133.1 | - | 1 | 35 | |||
AK216126.1 | - | 1 | 35 | |||
AK219241.1 | - | 1 | 35 | |||
BC138705.1 | - | 2 | 334 | 8861727 | ||
BC138706.1 | - | 2 | 336 | 8861728 | ||
BC158022.1 | - | 1 | 183 | 9007328 |