Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173414.3 | NP_775590.2 | 3, 4 | 1160 | ||
GenBank mRNA | AK031623.1 | - | 3, 4 | 1161 | ||
AK043841.1 | - | 3, 4 | 361 | |||
AK045254.1 | - | 3, 4 | 748 | |||
BC115944.1 | - | 2, 3 | 283 |