Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168507.1 | NP_001161979.1 | 3 | 965 | ||
NM_173453.3 | NP_775655.1 | 2 | 677 | |||
GenBank mRNA | AK077600.1 | - | 2 | 673 | ||
BC048514.1 | - | 2 | 425 | 6594842 |