Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130674.2 | NP_001124146.1 | 2 | 271 | ||
NM_001199671.1 | NP_001186600.1 | 3 | 1086 | |||
NM_001199672.1 | NP_001186601.1 | 3 | 1086 | |||
NM_001199673.1 | NP_001186602.1 | 2 | 271 | |||
NM_001219.4 | NP_001210.1 | 2 | 271 | |||
XM_017012659.1 | XP_016868148.1 | 2 | 192 | |||
GenBank mRNA | AF013759.1 | - | 2 | 177 | ||
AF257659.1 | - | 1 | 62 | |||
AF345637.1 | - | 2 | 164 | |||
AK027460.1 | - | 2 | 200 | |||
AK056338.1 | - | 3 | 974 | |||
AK074862.1 | - | 2 | 221 | |||
AK312446.1 | - | 2 | 214 | |||
BC013383.2 | - | 2 | 170 | 3507961 | ||
BT006826.1 | - | 1 | 119 | |||
BT020143.1 | - | 1 | 119 | |||
CR457039.1 | - | 1 | 119 | |||
CR541835.1 | - | 1 | 119 | |||
DA023987.1 | - | 2 | 271 | |||
DA703012.1 | - | 2 | 184 | |||
HM002604.1 | - | 3 | 1087 | |||
HM002605.1 | - | 3 | 1087 | |||
HM002606.1 | - | 2 | 271 | |||
HM002607.1 | - | 2 | 271 | |||
HM002609.1 | - | 2 | 271 | |||
HM002610.1 | - | 2 | 271 | |||
HM002611.1 | - | 2 | 271 | |||
HM002613.1 | - | 2 | 270 | |||
HM002615.1 | - | 3 | 1087 | |||
U67280.1 | - | 2 | 181 |