Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134791.2 | NP_001128263.1 | 16 | 1516 | ||
NM_133885.4 | NP_598646.4 | 19 | 1749 | |||
NM_173350.3 | NP_775485.2 | 17 | 1555 | |||
GenBank mRNA | AK035658.1 | - | 17 | 1504 | ||
AK075981.1 | - | 6 | 534 | |||
AK172010.1 | - | 17 | 1542 | |||
AK178534.1 | - | 2 | 65 | |||
AK202959.1 | - | 2 | 65 | |||
AK207117.1 | - | 2 | 65 | |||
BC021507.1 | - | 15 | 1377 | 5347121 | ||
BC023759.1 | - | 19 | 1704 | |||
BC026927.1 | - | 17 | 1438 | 5134780 | ||
BC089163.1 | - | 16 | 1457 | 5367603 |