Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173370.3 | NP_775546.2 | 13 | 2081 | ||
GenBank mRNA | AF533367.2 | - | 13 | 1990 | ||
BC026505.1 | - | 8 | 1248 | |||
BC043707.1 | - | 4 | 822 | |||
BC055292.1 | - | 13 | 2081 | 6491596 |