Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 3 | 1966 | ||
NM_173374.4 | NP_775550.2 | 4 | 1770 | |||
GenBank mRNA | AK018176.1 | - | 4 | 1439 | ||
AK027960.1 | - | 1 | 563 | |||
AK053674.1 | - | 4 | 1461 | |||
AK078715.1 | - | 3 | 1624 | |||
AK150535.1 | - | 4 | 1473 | |||
AK169203.1 | - | 4 | 1427 | |||
AK169623.1 | - | 4 | 1427 | |||
BC046773.1 | - | 4 | 1402 | 5708288 | ||
BC058627.1 | - | 4 | 1436 | 6401674 |