Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173765.3 | NP_776126.1 | 3 | 383 | ||
GenBank mRNA | AB095954.1 | - | 3 | 400 | ||
AK038779.1 | - | 4 | 393 | |||
AK043807.1 | - | 3 | 361 | |||
AK136849.1 | - | 3 | 381 | |||
AK148405.1 | - | 3 | 382 | |||
AK180643.1 | - | 284 | ||||
AK217387.1 | - | 3 | 333 | |||
BC058102.1 | - | 2 | 296 | |||
BC094507.1 | - | 2 | 160 | |||
BC128330.1 | - | 2 | 255 | 40110405 | ||
BY741513.1 | - | 3 | 384 |