Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173789.4 | NP_776150.2 | 3, 4 | 850 | ||
GenBank mRNA | AB098077.1 | - | 3, 4 | 301 | ||
AB120968.1 | - | 3, 4 | 850 | |||
AK044070.1 | - | 2, 3 | 401 | |||
BC103599.1 | - | 3, 4 | 735 | 40045925 | ||
BC103600.1 | - | 3, 4 | 735 | 40045926 | ||
BC103601.1 | - | 3, 4 | 735 | 40045923 | ||
DQ294234.1 | - | 3, 4 | 313 |