Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048192.2 | NP_001041657.1 | 2 | 202 | ||
NM_001294262.1 | NP_001281191.1 | 2 | 317 | |||
NM_174849.3 | NP_777274.1 | 2 | 206 | |||
GenBank mRNA | AK029676.1 | - | 2 | 207 | ||
AK034894.1 | - | 2 | 295 | |||
AK044195.1 | - | 1 | 1306 | |||
AK083656.1 | - | 2 | 253 | |||
BC057349.1 | - | 2 | 207 | 6813901 | ||
BY089079.1 | - | 2 | 275 | |||
BY100231.1 | - | 2 | 319 | |||
DQ867034.1 | - | 2 | 202 | |||
DQ867035.1 | - | 2 | 202 | |||
DQ867036.1 | - | 2 | 202 | |||
DQ867037.1 | - | 2 | 202 | |||
DQ867038.1 | - | 2 | 202 |