Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301174.1 | NP_001288103.1 | 2 | 151 | ||
NM_175024.4 | NP_778189.1 | 2 | 154 | |||
GenBank mRNA | AK080711.1 | - | 2 | 149 | ||
AK090312.1 | - | 2 | 152 | |||
BC100315.1 | - | 2 | 154 | 30931461 | ||
CK030777.1 | - | 2 | 135 | |||
DQ176852.1 | - | 2 | 141 |