Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyhin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175026.3 | NP_778191.1 | 2 | 542 | ||
GenBank mRNA | AK040884.1 | - | 2 | 313 | ||
AK080371.1 | - | 2 | 532 | |||
AK150817.1 | - | 2 | 433 | |||
BC096384.1 | - | 2 | 496 | 30752094 | ||
BC107378.1 | - | 2 | 376 | 40056703 | ||
BC107379.1 | - | 2 | 376 | 40056704 |
Gene Symbol: BC094916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024721.2 | NP_001019892.2 | 2 | 531 | ||
GenBank mRNA | AK156095.1 | - | 2 | 527 | ||
AK171847.1 | - | 2 | 522 | |||
BC094916.1 | - | 2 | 425 | 30074914 | ||
JN200820.1 | - | 1 | 225 |