Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175091.3 | NP_780300.2 | 6 | 1144 | ||
GenBank mRNA | AK048860.1 | - | 2 | 509 | ||
AK080913.1 | - | 6 | 1147 | |||
BC057370.1 | - | 6 | 1125 | 6835784 |