Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175116.4 | NP_780325.1 | 1 | 1266 | ||
GenBank mRNA | AK008952.1 | - | 1 | 1141 | ||
AK011967.1 | - | 1 | 1156 | |||
AK032901.1 | - | 1 | 1263 | |||
AK169602.1 | - | 1 | 1155 | |||
BC047931.1 | - | 1 | 1142 | 5251236 | ||
BC057175.1 | - | 1 | 1129 | |||
BC069991.1 | - | 1 | 1115 | 30244233 | ||
BC145940.1 | - | 1 | 892 | 40131243 | ||
BC145942.1 | - | 1 | 892 | 40131245 |